Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dengue Virus Type 2 NS4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532198
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532198 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532198 Supplier Invitrogen™ Supplier No. PA532198
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Dengue virus 2-infected C6/36, Dengue virus type 2 infected BHK-21, Dengue virus type 3 infected BHK-21. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Dengue fever and dengue hemorrhagic fever are acute febrile diseases, found in the tropics and Africa. Caused by one of four closely related virus serotypes of the genus Flavivirus, family Flaviviridae, each serotype is sufficiently different that there is no cross-protection and epidemics caused by multiple serotypes (hyperendemicity) can occur. Dengue is transmitted to humans by the Aedes aegypti (rarely Aedes albopictus) mosquito, which feeds during the day.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dengue Virus Type 2 NS4B
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene Alias Dengue NS4; Dengue NS4 protein
Host Species Rabbit
Immunogen Full length Dengue virus 2 NS4B Recombinant protein (Dengue virus 2 (strain 16681 PDK 53)).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.