Learn More
Invitrogen™ Dengue Virus Type 2 NS4B Polyclonal Antibody

Description
Recommended positive controls: Dengue virus 2-infected C6/36, Dengue virus type 2 infected BHK-21, Dengue virus type 3 infected BHK-21, Dengue virus type 2 infected huh7. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Dengue Virus Type 2 NS4B |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.01% thimerosal; pH 7 |
| Gene Alias | Dengue NS4; Dengue NS4 protein |
| Host Species | Rabbit |
| Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of Dengue virus NS4B protein (Dengue virus 2 (strain 16681 PDK 53)). The exact sequence is proprietary. |
| Purification Method | Antigen affinity chromatography |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.