Learn More
Invitrogen™ Dengue Virus Type 2 NS5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532200
Description
Recommended positive controls: Dengue virus type 1 infected BHK-21, Dengue virus type 2 infected BHK-21, Dengue virus type 3 infected BHK-21, Dengue virus type 4 infected BHK-21. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Dengue fever and dengue hemorrhagic fever are acute febrile diseases, found in the tropics and Africa. Caused by one of four closely related virus serotypes of the genus Flavivirus, family Flaviviridae, each serotype is sufficiently different that there is no cross-protection and epidemics caused by multiple serotypes (hyperendemicity) can occur. Dengue is transmitted to humans by the Aedes aegypti (rarely Aedes albopictus) mosquito, which feeds during the day.
Specifications
Dengue Virus Type 2 NS5 | |
Polyclonal | |
Unconjugated | |
Dengue NS5; Dengue NS5 protein | |
Recombinant protein encompassing a sequence within the C-terminus region of Dengue virus NS5 protein (Dengue virus 2 (strain 16681 PDK 53)). The exact sequence is proprietary. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.