Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DenLine Protection Plus Unisex 41 in. (104 cm) Long Closed Back Gown- Fluid Resistant Barrier (AAMI Level 2)
SDP

Catalog No. 22412106
Click to view available options
Color:
Royal Blue
White
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy

Catalog No. 22-412-106

Supplier: DenLine Protection Plus DL176LMROY

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Closed back gown provides AAMI Level 2 protection from incidental splash of blook, bodily fluids, and aerosols.

Primary Applications

· Anatomic Pathology · Clinical Pathology · Clinical Laboratory · Phlebotomy · Specimen Collection · Blood Collection, Testing, & Manufacturing · Human & Animal Research · Academia · Dialysis · Dental · Clean Room ISO 5/Class 100 - Medical Device, Pharma, Electronics

Wearer Protection Provided

  • One Piece Seamless front protects wearers from neck to below knees, arms, and covers tops of legs with no gaping. Textured Fabric holds fabric into position on top of legs when seated.
  • Adjustable Snaps at Neck for full Coverage. Back Ties at Waist
  • No patch pockets on front for splash of blood or OPIM to accumulate.
  • Closed Back Design offers protection to shoulders and back from splash traveling either side of employee’s face.
  • AAMI PB7- Level 2, Assists wearers comply with OSHA 29 CFR 1910:

Wearer Comfort

  • Overall Air Permeability 12 CFM (ASTM D737)
  • Raglan Sleeves and Loose Fit contribute to wearer comfort.

 Other Features

  • Hidden Side Seam Accessible Inside Pockets (2), protected from frontal splash.
  • Stain Resistant Fabric and Cuffs (blood, bleach and all aqueous reagents)
  • Anti-static Fabric prevents false instrument readings, damage to computers
  • Low-Lint Fabric prevents sample contamination, ideal for molecular testing
  • Badge Tag Loop
  • Durable Construction
    • Stainless Snap Closures at Back of Neck – Lifetime Guaranteed
    • Durable Cuffs for Overgloving – Bleach Safe all colors
  • Choice of 2 Colors, Royal and White, Sizes XS to 3XL
  • 225 Commercial Wash Tested; Low Lifetime Cost per Wash
  • Autoclavable Fabric (274°F/135°C).
  • Launder using Home/Laundromat or Commercial Equipment; Instructions furnished meet CDC/HICPAC Recommendations for processing medical bloodborne Linen

Specifications

Style Closed Back Gown
Construction 360° Fluid Resistance Front, Shoulders, and Back
For Use With (Application) Protection Against Incidental Splash of Blood & OPIM
Gender Unisex
Length (English) 41 in.
No. of Pockets None
Length (Metric) 104.1 cm
Product Type General Purpose Gown
Size Medium
Material Textured Polyester (99%), Carbon (1%)
Color Royal Blue
Closure Type Adjustable Snap at Neck, Back Ties at Waist
Disposable Reusable
Sterility Non-sterile
Cuff Style Knit
Finish Fluid and Stain Resistant, Anti-Static
Latex Free Yes
Certifications/Compliance OSHA 29 CFR 1910, AAMI PB70 Level 2
Weight (English) 0.6 lb.
Autoclavable Autoclavable
Permeability Overall Air Permeability 12 CFM (ASTM D737)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.