Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DenLine™ Uniforms Protection Plus™ DL171 Unisex Mid-Length Lab Coats
SDP

Catalog No. 22412040
Click to view available options
Color:
Black
Ceil Blue
Emerald Green
Royal Blue
White
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Mid-length lab coats provide AAMI PB70 Level 2 fluid resistance against incidental splash of blood, body fluids, aerosols, and all aqueous liquids. Suitable for cleanroom use.

  • Primary Applications:
    • Anatomic Pathology
    • Clinical Pathology
    • Clinical Laboratory
    • Phlebotomy
    • Specimen Collection
    • Blood Collection, Testing
    • Manufacturing
    • Human and Animal Research
    • Cleanroom ISO 5/Class 100 - Medical Device, Pharma, Electronics
  • High barrier protection against strike through from incidental splash of blood
  • Hydrostatic Pressure >32.5 CM (AATCC 127) and Impact Penetration <1.0 grams (AATCC 42) after completing 225 Hot Water Commercial Wash Cycles.
  • Complies with OSHA 29 CFR 1910
  • 360 degree protection front, shoulders, and back
  • Fluid resistant fabric, AAMI PB70 Level 2 Barrier
  • No cotton fabric used on shoulders or back as it would not meet OSHA 29 if splashed with blood or OPIM traveling rearward either side of the employee's head
  • Mid-Length 34 in. coat provides wearers 5 ft. 4 in. and shorter frontal coverage to knees
  • Provides all height wearers protection to ankles when worn in combination with DenLine style DL385 UltraLite Unisex Drawstring Pant
  • High air permeability (35 CFM) is used to make the back of coat for wearer comfort
  • Overall air permeability of coat front and back is 18 CFM (ASTM D737)
  • Stain fabric and cuffs (blood, bleach and ALL aqueous reagents and liquids)
  • Anti-static fabric prevents false instrument readings, damage to computers
  • Low lint fabric prevents sample contamination, ideal for molecular testing
  • Durable construction, 3 pockets, double stitched
  • Stainless snap closures
  • Durable cuffs for overgloving, bleach safe all colors
  • Hang loop outside back of collar
  • Side vents for pant pocket access
  • Choice of 5 Colors White, Ceil Blue, Royal, Black and Green, Sizes XS to 3XL

Specifications

Product Type General Purpose Lab Coat
Size X-Small
Gender Unisex
Material Polyester, Carbon
Color Emerald Green
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Static, Fluids
Style Mid-length
No. of Pockets 3
Description Durable Knit Cuffs for Overgloving, Bleach Safe Colors
Collar Type Traditional
Latex Free Yes
Size (English) Chest 30 to 32 in.
Size (English) Hip 33 to 35 in.
Size (Metric) Chest 76 to 81 cm
Size (Metric) Hip 84 to 89 cm
Certifications/Compliance OSHA, AAMI PB70 Level 2 Protection
For Use With (Application) Anatomic Pathology, Clinical Pathology, Clinical Laboratory, Phlebotomy, Specimen Collection, Research, Dental, Cleanroom ISO 5/Class 100
Length (English) 34 in.
Length (English) Cuff 4 in.
Length (English) Sleeve 35 in.
Length (Metric) 86 cm
Length (Metric) Cuff 10.1 cm
Autoclavable Autoclavable
Construction 360° Fluid Resistance Front, Shoulders, and Back
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.