Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DenLine™ Uniforms Protection Plus™ DL147 Unisex Short-Length Lab Jackets
SDP

Catalog No. 22412177
Click to view available options
Color:
Black
Royal Blue
White
Size:
2X-Large
4X-Large
5X-Large
This item is not returnable. View return policy

Catalog No. 22-412-177

Supplier: DenLine™ Uniforms DL1474XLROY

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Short-length lab coats provide AAMI PB70 Level 2 fluid resistance against incidental splash of blood, body fluids, aerosols, and all aqueous liquids. Suitable for cleanroom use.

  • Primary Applications:
    • Anatomic Pathology
    • Clinical Pathology
    • Clinical Laboratory
    • Phlebotomy
    • Specimen Collection
    • Blood Collection, Testing
    • Manufacturing
    • Human and Animal Research
    • Cleanroom ISO 5/Class 100 - Medical Device, Pharma, Electronics
  • High barrier protection against strike through from incidental splash of blood
  • Hydrostatic Pressure >32.5 CM (AATCC 127) and Impact Penetration <1.0 grams (AATCC 42) after completing 225 Hot Water Commercial Wash Cycles.
  • Complies with OSHA 29 CFR 1910
  • 360 degree protection front, shoulders, and back
  • Fluid resistant fabric, AAMI PB70 Level 2 Barrier
  • No cotton fabric used on shoulders or back as it would not meet OSHA 29 if splashed with blood or OPIM traveling rearward either side of the employee's head
  • Short-length 31.5 in.
  • Provides all height wearers protection to ankles when worn in combination with DenLine style DL385 UltraLite Unisex Drawstring Pant
  • High air permeability (35 CFM) is used to make the back of coat for wearer comfort
  • Overall air permeability of coat front and back is 18 CFM (ASTM D737)
  • Stain fabric and cuffs (blood, bleach and ALL aqueous reagents and liquids)
  • Anti-static fabric prevents false instrument readings, damage to computers
  • Low lint fabric prevents sample contamination, ideal for molecular testing
  • Durable construction, 3 pockets, double stitched
  • Stainless snap closures
  • Durable cuffs for overgloving, bleach safe all colors
  • Hang loop outside back of collar
  • Side vents for pant pocket access

Order Info

Colors: white, royal and black. Sizes available: XS to 5XL.

Specifications

Product Type General Purpose Lab Coat
Size 4X-Large
Gender Unisex
Material Polyester
Color Royal Blue
Closure Type Snaps
Disposable Reusable
Sterility Non-sterile
Resistance Fluids, Stains
Style Short
No. of Pockets 3
Collar Type No Collar
Sleeve Style Raglan
Size (English) Chest 58 to 60 in.
Size (English) Hip 54 to 57.5 in.
Certifications/Compliance AAMI PB70 Level 2
For Use With (Application) Dental, Blood Collection, Nursing, Quasi-Cleanroom Applications
Item Description EZ Snap
Length (English) 31.5 in.
Length (English) Cuff 4 in.
Length (Metric) 80 cm
Length (Metric) Cuff 10.16 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.