Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DENN/MADD Domain Containing 2D Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP230995
Description
DENN/MADD Domain Containing 2D Polyclonal specifically detects DENN/MADD Domain Containing 2D in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DENN/MADD Domain Containing 2D | |
Polyclonal | |
Western Blot 0.4 ug/ml, Simple Western 1:25, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Q9H6A0 | |
DENND2D | |
This antibody was developed against a recombinant protein corresponding to amino acids: PPQDNSGEALKEPERAQEHSLPNFAGGQHFFEYLLVVSLKKKRSEDDYEPIITYQFPKRENLLRGQQEEEERLLKAI | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
DENN Domain-Containing Protein 2D, DENND2D, RP5-1180E21.2 | |
Rabbit | |
Affinity Purified | |
RUO | |
79961 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction