Learn More
Invitrogen™ DENND1B Polyclonal Antibody

Description
This antibody is predicted to react with mouse based on sequence homology.
Specifications
Specifications
| Antigen | DENND1B |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | DENND1B |
| Gene Accession No. | Q6P3S1 |
| Gene Alias | 4632404N19Rik; 4930467M19Rik; 6820401H01Rik; ASL1/9AS1-1 fusion; C1ORF18; C1orf218; Connecdenn 2; DENN domain containing 1B; DENN domain-containing protein 1B; DENN/MADD domain containing 1B; Dennd1b; F730008N07Rik; Fam31b; family with sequence similarity 31, member B; Protein FAM31B; RP11-53I24.2 |
| Gene Symbols | DENND1B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.