Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DENND1C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560144
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560144 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560144 Supplier Invitrogen™ Supplier No. PA560144
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PLSPEDEGCP WAEEALDSSF LGSGEELDLL SEILDSLSMG AKSAGSLRPS QSLDCCHRGD LDSCFSLPNI PRWQPDDKKL Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 61%.

Clathrin-mediated endocytosis is a major mechanism for internalization of proteins and lipids. Members of the connecdenn family, such as DENND1C, function as guanine nucleotide exchange factors (GEFs) for the early endosomal small GTPase RAB35 and bind to clathrin and clathrin adaptor protein-2. Thus, connecdenns link RAB35 activation with the clathrin machinery.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DENND1C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DENND1C
Gene Accession No. Q8IV53
Gene Alias Connecdenn 3; DENN domain containing 1C; DENN domain-containing protein 1C; DENN/MADD domain containing 1C; DENND1C; FAM31C; family with sequence similarity 31, member C; Protein FAM31C
Gene Symbols DENND1C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DENND1C. Recombinant protein control fragment (Product #RP-99239).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79958
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.