Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DENND1C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113099
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113099 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113099 Supplier Invitrogen™ Supplier No. PA5113099
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DENND1C Polyclonal Antibody for Western Blot, IHC (P), ELISA

Clathrin-mediated endocytosis is a major mechanism for internalization of proteins and lipids. Members of the connecdenn family, such as DENND1C, function as guanine nucleotide exchange factors (GEFs) for the early endosomal small GTPase RAB35 and bind to clathrin and clathrin adaptor protein-2. Thus, connecdenns link RAB35 activation with the clathrin machinery.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DENND1C
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene DENND1C
Gene Accession No. Q8IV53
Gene Alias Connecdenn 3; DENN domain containing 1C; DENN domain-containing protein 1C; DENN/MADD domain containing 1C; DENND1C; FAM31C; family with sequence similarity 31, member C; Protein FAM31C
Gene Symbols DENND1C
Host Species Rabbit
Immunogen Recombinant Human DENN domain-containing protein 1C protein (601-801AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 363326, 79958
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.