Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DENND2C Polyclonal Antibody
Supplier: Thermo Scientific PA5112989
Description

Specifications
DENND2C | |
Polyclonal | |
Unconjugated | |
DENND2C | |
A930010I20Rik; DENN domain containing 2C; DENN domain-containing protein 2C; DENN/MADD domain containing 2C; DENND2C; dJ1156J9.1; RGD1308197; RP5-1156J9.1 | |
Rabbit | |
Protein G | |
RUO | |
163259 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
4 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q68D51 | |
DENND2C | |
Recombinant Human DENN domain-containing protein 2C protein (1-300AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction