Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DENND2C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563042
Description
Immunogen sequence: WEGRANGISN PEKWCPKDFG VRYNCHQEIR LKKNPIAERK SKNLDVTSRE NVGLDINENT KSHDQSENEN KKHEYDDTHF FKNESESNWV CSRVKEIESC KED Highest antigen sequence identity to the following orthologs: Mouse - 44%, Rat - 47%.
Guanine nucleotide exchange factor (GEF) which may activate RAB9A and RAB9B. Promotes the exchange of GDP to GTP, converting inactive GDP-bound Rab proteins into their active GTP-bound form.
Specifications
DENND2C | |
Polyclonal | |
Unconjugated | |
DENND2C | |
A930010I20Rik; DENN domain containing 2C; DENN domain-containing protein 2C; DENN/MADD domain containing 2C; DENND2C; dJ1156J9.1; RGD1308197; RP5-1156J9.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
163259 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q68D51 | |
DENND2C | |
Recombinant protein corresponding to Human DENND2C. Recombinant protein control fragment (Product #RP-104237). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction