Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DENND2C Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS9667R

Description
Guanine nucleotide exchange factor (GEF) which may activate RAB9A and RAB9B. Promotes the exchange of GDP to GTP, converting inactive GDP-bound Rab proteins into their active GTP-bound form.Specifications
DENND2C | |
Polyclonal | |
Unconjugated | |
DENND2C | |
A930010I20Rik; BOS_3083; DENN domain containing 2C; DENN domain-containing protein 2C; DENN/MADD domain containing 2C; DENND2C; dJ1156J9.1; RGD1308197; RP5-1156J9.1 | |
Rabbit | |
Protein A | |
RUO | |
163259 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q68D51 | |
DENND2C | |
KLH conjugated synthetic peptide derived from human DENND2C. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction