Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DENND3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564996
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564996 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564996 Supplier Invitrogen™ Supplier No. PA564996
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PTSFLMGCHLD HFEEVSKEAD GLVLINIDHG SITYSKSTDD NVDIPDVPLL AAQTFIQRVQ SLQLHHELHA AHLLSSTDLK EGRAHRRSWQ QKLNCQIQQT T Highest antigen sequence identity to the following orthologs - mouse 81%, rat 78%.

Guanine nucleotide exchange factor (GEF) activating RAB12. Promotes the exchange of GDP to GTP, converting inactive GDP-bound RAB12 into its active GTP-bound form (PubMed:20937701). Regulates autophagy in response to starvation through RAB12 activation. Starvation leads to ULK1/2-dependent phosphorylation of Ser-472 and Ser-490, which in turn allows recruitment of 14-3-3 adapter proteins and leads to up-regulation of GEF activity towards RAB12. Also plays a role in protein transport from recycling endosomes to lysosomes, regulating, for instance, the degradation of the transferrin receptor and of the amino acid transporter PAT4 (PubMed:20937701). Starvation also induces phosphorylation at Tyr-858, which leads to up-regulated GEF activity and initiates autophagy. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DENND3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DENND3
Gene Accession No. A2RUS2
Gene Alias AI447457; DENN domain containing 3; DENN domain-containing protein 3; DENN/MADD domain containing 3; Dennd3; E030003N15Rik; KIAA0870
Gene Symbols DENND3
Host Species Rabbit
Immunogen Recombinant Human DENND3. Recombinant protein control fragment (Product #RP-105756).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22898
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.