Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DENND4A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564219
Description
Immunogen sequence: LDTSALSVQG NFDLNSKSKL QENFCTRSIQ IPANRSKTAM SKCPIFPMAR SISTSGPLDK EDTGRQKLIS TGSLPATLQG ATDSLGL Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 82%.
Probable guanine nucleotide exchange factor (GEF) which may activate RAB10. Promotes the exchange of GDP to GTP, converting inactive GDP-bound Rab proteins into their active GTP-bound form. According to PubMed:8056341, it may bind to ISRE-like element (interferon-stimulated response element) of MYC P2 promoter.
Specifications
DENND4A | |
Polyclonal | |
Unconjugated | |
DENND4A | |
AI115600; AI851943; c-myc promoter binding protein; C-myc promoter-binding protein; DENN domain containing 4A; DENN domain-containing protein 4A; DENN/MADD domain containing 4A; DENND4A; F730015K02Rik; IRLB; MYCPBP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10260 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z401 | |
DENND4A | |
Recombinant protein corresponding to Human DENND4A. Recombinant protein control fragment (Product #RP-104257). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction