Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DENND4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103751
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103751 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103751 Supplier Invitrogen™ Supplier No. PA5103751
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DENND4A.

Probable guanine nucleotide exchange factor (GEF) which may activate RAB10. Promotes the exchange of GDP to GTP, converting inactive GDP-bound Rab proteins into their active GTP-bound form. According to PubMed:8056341, it may bind to ISRE-like element (interferon-stimulated response element) of MYC P2 promoter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DENND4A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DENND4A
Gene Accession No. Q7Z401
Gene Alias AI115600; AI851943; c-myc promoter binding protein; C-myc promoter-binding protein; DENN domain containing 4A; DENN domain-containing protein 4A; DENN/MADD domain containing 4A; DENND4A; F730015K02Rik; IRLB; MYCPBP
Gene Symbols DENND4A
Host Species Rabbit
Immunogen A synthesized peptide derived from human DENND4A(Accession Q7Z401), corresponding to amino acid residues L989-L1039.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102442, 10260
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.