Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DENND4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584858
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584858 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584858 Supplier Invitrogen™ Supplier No. PA584858
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSPWLTPDPA SVQVRLLWDV LTPDPNSCPP LYVLWRVHSQ IPQRVVWPGP VPASLSLALL ESVLRHVGLN EVHKAVGLLL ETLG Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 93%.

DENND4B is a protein coding gene. Guanine nucleotide exchange factor (GEF) which may activate RAB10. Promotes the exchange of GDP to GTP, converting inactive GDP-bound Rab proteins into their active GTP-bound form.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DENND4B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DENND4B
Gene Accession No. O75064
Gene Alias Brain specific protein 4; brain-specific gene 4 protein; Bsg4; Bsp4; DENN domain containing 4B; DENN domain-containing protein 4B; DENN/MADD domain containing 4B; DENND4B; KIAA0476; RGD1560320
Gene Symbols DENND4B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DENND4B. Recombinant protein control fragment (Product #RP-108579).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9909
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.