Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DENND4C Monoclonal Antibody (3E8), FITC
GREENER_CHOICE

Catalog No. PIMA545961
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545961 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545961 Supplier Invitrogen™ Supplier No. MA545961
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

A 1:1,000 dilution of MA5-45961 was sufficient for detection of DENND4C in A549, MEF cells by Western Blot analysis. Detects approximately 213 kDa.

DENND4C is guanine nucleotide exchange factor (GEF) activating RAB10. It promotes the exchange of GDP to GTP, converting inactive GDP-bound RAB10 into its active GTP-bound form. Thereby, stimulates SLC2A4/GLUT4 glucose transporter-enriched vesicles delivery to the plasma membrane in response to insulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DENND4C
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3E8
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
Gene DENND4C
Gene Accession No. A6H8H2, Q5VZ89
Gene Alias 1700065A05Rik; AA420392; bA513M16.3; C9orf55; C9orf55B; DENN domain containing 4C; DENN domain-containing protein 4C; DENN domain-containing protein 4C-like protein; DENN/MADD domain containing 4A pseudogene; DENN/MADD domain containing 4C; Dennd4c; likely ortholog of H. sapiens chromosome 9 open reading frame 55 (C9orf55); RAB10GEF; RGD1308489
Gene Symbols DENND4C
Host Species Mouse
Immunogen Full Length Human Recombinant Dennd4c protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 313340, 329877, 55667
Target Species Human, Mouse, Rat
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.