Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DENND4C Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142659
Description
Additional Information: This antibody is expected to recognize both reported isoforms (NP_908976.1; NP_001074483.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.
DENND4C is guanine nucleotide exchange factor (GEF) activating RAB10. It promotes the exchange of GDP to GTP, converting inactive GDP-bound RAB10 into its active GTP-bound form. Thereby, stimulates SLC2A4/GLUT4 glucose transporter-enriched vesicles delivery to the plasma membrane in response to insulin.
Specifications
| DENND4C | |
| Polyclonal | |
| Unconjugated | |
| DENND4C | |
| 1700065A05Rik; AA420392; bA513M16.3; C9orf55; C9orf55B; DENN domain containing 4C; DENN domain-containing protein 4C; DENN domain-containing protein 4C-like protein; DENN/MADD domain containing 4A pseudogene; DENN/MADD domain containing 4C; Dennd4c; likely ortholog of H. sapiens chromosome 9 open reading frame 55 (C9orf55); RAB10GEF; RGD1308489 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 329877 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| A6H8H2 | |
| DENND4C | |
| Peptide with sequence C-RFKPQSPYRAYKDRS. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction