Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DENND4C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546141
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546141 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546141 Supplier Invitrogen™ Supplier No. PA546141
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KHNQIITEET GSAVEPSDEI KRASGDVQTM KISSVPNSLS KRNVSLTRSH Sequence homology: Cow: 85%; Dog: 86%; Guinea Pig: 79%; Horse: 85%; Human: 100%; Rabbit: 83%.

DENND4C is guanine nucleotide exchange factor (GEF) activating RAB10. It promotes the exchange of GDP to GTP, converting inactive GDP-bound RAB10 into its active GTP-bound form. Thereby, stimulates SLC2A4/GLUT4 glucose transporter-enriched vesicles delivery to the plasma membrane in response to insulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DENND4C
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DENND4C
Gene Accession No. Q5VZ89
Gene Alias 1700065A05Rik; AA420392; bA513M16.3; C9orf55; C9orf55B; DENN domain containing 4C; DENN domain-containing protein 4C; DENN domain-containing protein 4C-like protein; DENN/MADD domain containing 4A pseudogene; DENN/MADD domain containing 4C; Dennd4c; likely ortholog of H. sapiens chromosome 9 open reading frame 55 (C9orf55); RAB10GEF; RGD1308489
Gene Symbols DENND4C
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human DENND4C (aa 1590-1639).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55667
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.