Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DENND5A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562705
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562705 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562705 Supplier Invitrogen™ Supplier No. PA562705
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MHNAEYDVLH APPADDRDQS SMEDGEDTPV TKLQR Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 80%.

DENND5A (DENN domain-containing protein 5A) is a guanine nucleotide exchange factor (GEF) which activates RAB6A and RAB39A/RAB39B. It promotes the exchange of GDP to GTP by converting inactive GDP-bound Rab proteins to their active GTP form. DENND5A is also involved in the negative regulation of neurtie outgrowth. Mutations affecting this gene can result in epileptic encephalopathy, early infantile, 49 (EIEE49).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DENND5A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DENND5A
Gene Accession No. Q6IQ26
Gene Alias 1500012B19Rik; AJ245569; AW496491; DENN domain containing 5A; DENN domain-containing protein 5A; DENN/MADD domain containing 5A; DENND5A; KIAA1091; mKIAA1091; ORF37; Rab6 interacting protein 1; rab6-interacting protein 1; RAB6IP1
Gene Symbols DENND5A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DENND5A. Recombinant protein control fragment (Product #RP-100833).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23258
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.