Learn More
Invitrogen™ DENND6B Polyclonal Antibody

Description
Immunogen sequence: HFDGWYRQRH KEMALKLEAL HLEAICEANI ETWMKDKSEV EVVDLVLKLR EKLVRAQGHQ LPVKEATLQR AQLYIETVIG SLPKDLQAVL CPP Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
Specifications
Specifications
| Antigen | DENND6B |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | DENND6B |
| Gene Accession No. | Q8NEG7 |
| Gene Alias | 1700027J05Rik; AFI1B; AI414881; DENN domain containing 6B; DENN domain-containing protein 6B; DENN/MADD domain containing 6B; Dennd6b; FAM116B; family with sequence similarity 116, member B; hypothetical protein MGC38041; protein DENND6B; protein FAM116B; RGD1308012 |
| Gene Symbols | DENND6B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.