Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DENR Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP150042AF350
Description
DENR Polyclonal antibody specifically detects DENR in Human, Mouse samples. It is validated for Western BlotSpecifications
DENR | |
Polyclonal | |
Western Blot | |
density-regulated protein, DRP1Protein DRP1, DRPsmooth muscle cell associated protein-3, SMAP-3, Smooth muscle cell-associated protein 3 | |
The immunogen for this product maps to a region between residue 148 and 198 of human Density-Regulated Protein using the numbering given in entry NP_003668.2 (GeneID 8562). | |
0.1 mL | |
Signal Transduction | |
8562 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction