Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DENR Monoclonal Antibody (1542CT106.51.79)
GREENER_CHOICE

Catalog No. PIMA524740
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524740 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524740 Supplier Invitrogen™ Supplier No. MA524740
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DENR Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

This gene encodes a protein whose expression was found to increase in cultured cells at high density but not during growth arrest. This gene was also shown to have increased expression in cells overexpressing HER-2/neu proto-oncogene. The protein contains an SUI1 domain. In budding yeast, SUI1 is a translation initiation factor that along with eIF-2 and the initiator tRNA-Met, directs the ribosome to the proper translation start site. Proteins similar to SUI have been found in mammals, insects, and plants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DENR
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1542CT106.51.79
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene DENR
Gene Accession No. O43583
Gene Alias 1500003K04Rik; Denr; density regulated re-initiation and release factor; density-regulated protein; DRP; DRP1; H14; Protein DRP1; SMAP-3; smooth muscle cell associated protein-3; smooth muscle cell-associated protein 3
Gene Symbols DENR
Host Species Mouse
Immunogen human DENR recombinant protein.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8562
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.