Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DENR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554507
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554507 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554507 Supplier Invitrogen™ Supplier No. PA554507
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AADISESSGA DCKGDPRNSA KLDADYPLRV LYCGVCSLPT EYCEYMPDVA KCRQWLEKNF PNEFAKLTVE NSPKQEAGIS EGQGTAGEEE EKK Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 86%.

This gene encodes a protein whose expression was found to increase in cultured cells at high density but not during growth arrest. This gene was also shown to have increased expression in cells overexpressing HER-2/neu proto-oncogene. The protein contains an SUI1 domain. In budding yeast, SUI1 is a translation initiation factor that along with eIF-2 and the initiator tRNA-Met, directs the ribosome to the proper translation start site. Proteins similar to SUI have been found in mammals, insects, and plants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DENR
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DENR
Gene Accession No. O43583
Gene Alias 1500003K04Rik; Denr; density regulated re-initiation and release factor; density-regulated protein; DRP; DRP1; H14; Protein DRP1; SMAP-3; smooth muscle cell associated protein-3; smooth muscle cell-associated protein 3
Gene Symbols DENR
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DENR. Recombinant protein control fragment (Product #RP-92545).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8562
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.