Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Density Ball
SDPSCI_ED

Catalog No. S96824
Click to view available options
Material:
Metal

Demonstrate the effects of temperature and mineral content on density

Metal ball floats in cold water and sinks in hot.

  • Can be used density effects of ocean currents by varying salt content as well as temperature

Specifications

For Use With (Application) To demonstrate the effects of temperature and mineral content on density
Product Type Density Ball
Material Metal
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.