Learn More
United Scientific Supplies Density Cube Sets
SDPSCI_ED
Catalog No. S23980
Click to view available options
Quantity:
10 Cubes
12 Cubes
4 Cubes
Demonstrate a variety of densities with equal shaped cubes. The United Scientific Supplies Density Cube Sets include one of each of a variety of cubes for density studies.
  • For use in science experiments involving density
  • Cube measurements are equal
Specifications
Product Type Density Cubes
Description Polybag
Includes One each of acrylic, pine, nylon, and aluminum cubes
Quantity 4 Cubes
Height (English) 1 in.
Width (English) 1 in.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.