Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DENTT Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501566675
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-6675 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-6675 Supplier Bethyl Laboratories, Inc Supplier No. A304012A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

This antibody recognizes human DENTT (Differentially Expressed Nucleolar TGF-Beta1 Target) protein, a gene product expressed in several normal tissues with the highest level of expression in brain. DENTT is involved in responses mediated by TGF-Beta1 and has distinct domains for direction to different subnuclear locations.

Specifications

Antigen DENTT
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene TSPYL2
Gene Accession No. Q9H2G4
Gene Alias RP1-290F12.2, CDA1, CINAP, CTCL, DENTT, HRIHFB2216, NP79, SE204, TSPX
Gene Symbols TSPYL2
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64061
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.