Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DENTT Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS7051R

Description
This gene encodes the beta-subunit of glucosidase II, an N-linked glycan-processing enzyme in the endoplasmic reticulum (ER). This protein is an acidic phospho-protein known to be a substrate for protein kinase C. Mutations in this gene have been associated with the autosomal dominant polycystic liver disease (PCLD). Alternatively spliced transcript variants encoding distinct isoforms have been observed.Specifications
DENTT | |
Polyclonal | |
Unconjugated | |
TSPYL2 | |
CASK interacting nucleosome assembly protein; CASK-interacting nucleosome assembly protein; CDA1; Cell division autoantigen 1; cell division autoantigen 1 nucleolar protein; Cinap; CTCL; CTCL tumor antigen se20-4; CTCL-associated antigen se20-4; Cutaneous T-cell lymphoma-associated antigen se20-4; cutaneous T-cell lymphoma-associated tumor antigen se20-4; Dentt; differentially expressed nucleolar TGF-beta1 target; Differentially-expressed nucleolar TGF-beta1 target protein; DXBwg1396e; DXHXS1008E; E130307F10Rik; HRIHFB2216; NP79; Nuclear protein of 79 kDa; nucleolar TGF-beta1 target protein; RGD1563283; RP1-290F12.2; SE204; testis-specific protein Y encoded-like 2; Testis-specific Y-encoded-like protein 2; Tspx; TSPY like 2; Tspyl2; TSPY-like 2; TSPY-like protein 2 | |
Rabbit | |
Protein A | |
RUO | |
64061 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9H2G4 | |
TSPYL2 | |
KLH conjugated synthetic peptide derived from human DENTT. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction