Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ deoD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598329
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598329 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598329 Supplier Invitrogen™ Supplier No. PA598329
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

deoD Polyclonal Antibody for Western Blot, ELISA

Cleavage of guanosine or inosine to respective bases and sugar-1-phosphate molecules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen deoD
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Alias PNP; PNP-DeoD; Purine nucleoside phosphorylase DeoD
Host Species Rabbit
Immunogen Recombinant Escherichia coli Purine nucleoside phosphorylase DeoD-type protein (2-239AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.