Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deoxybenzoin, 97%

Catalog No. p-99202
Click to view available options
Quantity:
25 g
100 g

CAS: 451-40-1 | C14H12O | 196.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 451-40-1
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.25
MDL Number MFCD00003081
InChI Key OTKCEEWUXHVZQI-UHFFFAOYSA-N
Synonym 2-phenylacetophenone, deoxybenzoin, benzyl phenyl ketone, ethanone, 1,2-diphenyl, desoxybenzoin, 1,2-diphenylethan-1-one, phenyl benzyl ketone, benzoin, deoxy, acetophenone, 2-phenyl, deoxy benzoin
PubChem CID 9948
IUPAC Name 1,2-diphenylethanone
SMILES O=C(CC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 52.0°C to 57.0°C
Color White
Boiling Point 320.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5CH2COC6H5
Quantity 25 g
Beilstein 07, II, 368
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in alcohols and ketones
Formula Weight 196.25
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Deoxybenzoin, 98%
Show More Show Less
EINECSNumber 207-193-2
RTECSNumber AM9662500
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.