Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Deoxycholic Acid, Sodium Salt
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
25 g
100 g
500 g
Description
- Deoxycholic acid, sodium salt is an anionic detergent useful for the extraction of membrane receptors and other plasma membrane proteins and for nuclei isolation.
- Deoxycholic acid has been used in a modified procedure to recover 40 to 80% of a protein from a 1 μg/mL solution.

Specifications
Specifications
Color | White |
Product Type | Detergent |
CAS | 302-95-4 |
Molecular Formula | C24H39NaO4 |
Formula Weight | 414.55 |
MDL Number | MFCD00064139 |
Form | Powder |
InChI Key | FHHPUSMSKHSNKW-SMOYURAASA-M |
SMILES | [Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C |
IUPAC Name | sodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate |
Show More |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction