Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Deoxycholic Acid, Sodium Salt, ≥96%, Calbiochem™,

Catalog No. 26410125GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
26-410-125GM 25 g Glass Bottle
26-410-11KG 1 kg Glass Bottle
26-410-1100GM 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 26-410-125GM Supplier MilliporeSigma™ Supplier No. 26410125GM

Chemical Identifiers

CAS 302-95-4
Molecular Formula C24H39NaO4
Molecular Weight (g/mol) 414.56
MDL Number MFCD00064139
InChI Key FHHPUSMSKHSNKW-SMOYURAASA-M
PubChem CID 91810855
IUPAC Name sodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate
SMILES [Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C

Specifications

Color White
Quantity 25 g
Packaging Glass Bottle
Percent Purity ≥96% (TLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.