Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Deoxycytidylate deaminase Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299193DL594
Description
Deoxycytidylate deaminase Polyclonal specifically detects Deoxycytidylate deaminase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Deoxycytidylate deaminase | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
dCMP deaminaseEC 3.5.4.12, deoxycytidylate deaminase, MGC111062 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Deoxycytidylate deaminase. | |
0.1 mL | |
Lipid and Metabolism | |
1635 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction