Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Deoxyribonuclease I (Bovine), Molecular Biology Grade

Catalog No. 89941799
Click to view available options
Quantity:
100 U

Used for Enzyme

#REF!

Specifications

Accession Number P00639
Concentration ∼ 1 mg/mL
For Use With (Application) Enzyme Activity
Formulation In 50% glycerol and 1mM calcium chloride
Gene ID (Entrez) 282217
Molecular Weight (g/mol) 31.3kDa
Name Deoxyribonuclease I (Bovine), Molecular Biology Grade
Preparation Method Native protein chromatographically purified from Bovine Pancreas
Quantity 100 U
Storage Requirements Store at -20°C.
Regulatory Status RUO
Common Name DNASE1
Gene Symbol DNASE1
Assays Deoxyribonuclease I Assay; Nick translation; DNA mapping; Isolation of nuclear RNA and protein; Plasmid construction; RNA polymerase synthesis of RNA probes and RT-PCR.
Biological Activity ≥2,000 Kunitz units per ml. One Unit causes an increase in absorbance at 260nm of 0.001 per minute per ml at 25°C when acting upon highly polymerized DNA at pH 5.0.
Species Bovine
Protein Tag None
Expression System Native protein chromatographically purified from Bovine Pancreas
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.