Learn More
Thermo Scientific Chemicals Deoxyribonuclease I, bovine pancreas
Description
Deoxyribonuclease I is used for the removal of DNA from protein samples. DNAse I is used to nick DNA as a first step to incorporate labeled bases into DNA. The enzyme from Sigma has been used during the isolation of plasma membrane vesicles from Neurospora crassa cell culture.1 It has also been used along with trypsin for the preparation of single cell suspension from rat testes. Deoxyribonuclease I from bovine pancreas has been used in a study to compare several procedures for reducing RNase contamination in preparations of DNase. Deoxyribonuclease I from bovine pancreas has also been used in a study to investigate the effect of the composition of sodium dodecyl sulfate preparations on the renaturation of enzymes after polyacrylamide gel electrophoresis
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Deoxyribonuclease I is an endonuclease that acts on phosphodiester bonds adjacent to pyrimidines to produce polynucleotides with terminal 5′-phosphates. In the presence of Mg2+, this compound cleaves each strand of DNA independently
Application
- In biomedical research, this compound is used to nick DNA as a first step to integrate labeled bases into DNA
- It can be used to eliminate genomic DNA from RNA solutions before RT-PCR
- It is also used during isolation and purification of plasma membrane vesicles from Neurospora crassa cell culture
- It can decrease viscosity in cell culture lysates, providing better yields in primary cell isolation
- It is also commonly used in combination with trypsin for the preparation of single-cell suspension from rats
- Deoxyribonuclease I, bovine pancreas has been used in some studies in order to compare different methods for reducing RNase contamination in preparations of DNase
- This compound can be applied in studies to investigate the impact of the composition of sodium dodecyl sulfate (SDS) preparations on the renaturation of enzymes after polyacrylamide gel electrophoresis
Specifications
Specifications
| Quantity | 1 g |
| Form | Lyophilized |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.