Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deoxyribonuclease I, MP Biomedicals™

Catalog No. ICN10057580 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10mg
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10057580 0.1g
ICN10057510 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10057580 Supplier MP Biomedicals, Inc Supplier No. 0210057580
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 9003-98-9

Specifications

Color White
Quantity 0.1g
Product Type Deoxyribonuclease I
Form Powder
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.