Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deoxyribonuclease I Polyclonal Antibody, Rockland™
SDP

Catalog No. 501991038
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1038 25 μL
50-199-1037 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1038 Supplier Rockland Immunochemicals Supplier No. 2004161S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-DEOXYRIBONUCLEASE I is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum as well as purified and partially purified Deoxyribonuclease A [Bovine Pancreas]. Cross reactivity against Deoxyribonuclease A from other tissues and species may occur but have not been specifically determined.

Internucleosomal DNA fragmentation following the activation of endonucleases is the common end point of apoptosis. DNase I, a Ca2+/Mg2+-dependent endonuclease ubiquitously expressed in mammalian tissues, has been implicated to mediate internucleosomal DNA degradation in human cells undergoing apoptosis. DNase I is highly polymorphic, and at least six alleles of DNase I are known. DNase II, the ubiquitously expressed acidic deoxyribonuclease, acts downstream of caspase activation and may also induce DNA digestion during apoptosis. DNase I cleaves DNA to 5'-phosphodinucleotide and 5'-phospho- oligonucleotide end-products, whereas DNase II cleaves DNA to 3'-phosphomononucleotide and 3'-phosphooligonucleotide end-products. The mechanism by which DNase II cuts DNA is similar to DNase I, which produces nicks rather than double-strand cuts. DNase II is usually present in cytoplasm of epithelial cells, but it appears concentrated in the nuclei of lens fibers. In contrast, DNase I is always concentrated in nuclei of epithelial and fiber cells.

Specifications

Antigen Deoxyribonuclease I
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene DNASE1
Gene Accession No. P00639
Gene Alias AI788650; Deoxyribonuclease 1; Deoxyribonuclease I; Deoxyribonuclease1; Deoxyribonuclease-1; DeoxyribonucleaseI; DNASE 1; DNase I; DNase I lysosomal; DNase I, lysosomal; DNASE1; DNaseI; DNL 1; Dnl1; Dornase alfa; DRNI; FLJ38093; human urine deoxyribonuclease I
Gene Symbols DNASE1
Host Species Rabbit
Immunogen Deoxyribonuclease A [Bovine Pancreas]
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282217
Target Species Bovine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.