Learn More
Deoxyribonucleoside Triphosphate Mixture, [α-32P] , MP Biomedical
Catalog No. MP133030H01 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 Vial
  • Deoxyadenosine-5´-Triphosphate, [α-32P]
  • Deoxycytidine-5´-Triphosphate, [α-32P]
  • Deoxyguanosine-5´-Triphosphate, [α-32P]
  • Thymidine-5´-Triphosphate, [α-32P]
  • 125μCi, 4.62 MBq of each mixed in one vial
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Vial
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.