Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DEP-1/CD148 Antibody (MEM-CD148/05) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP221790
Description
DEP-1/CD148 Monoclonal specifically detects DEP-1/CD148 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
DEP-1/CD148 | |
Monoclonal | |
1.0 mg/mL | |
Flow Cytometry 1 to 5 μg/mL, CyTOF-ready | |
PTPRJ | |
Human recombinant CD148 (amino acids 1-444). | |
Protein A purified | |
RUO | |
Primary | |
The mouse monoclonal antibody MEM-CD148/05 recognizes CD148, a highly glycosylated up to 250 kDa receptor-like protein tyrosin phosphatase expressed mainly in lymphocytes, myeloid cells and epithelial cells. | |
Store at 4C. Do not freeze. | |
IgG2b |
Flow Cytometry, CyTOF | |
MEM-CD148/05 | |
Unconjugated | |
CD148, CD148 antigen, Density-enhanced phosphatase 1, DEP-1, DEP1EC 3.1.3.48, HPTP eta, HPTPeta, human density enhanced phosphatase-1, protein tyrosine phosphatase, receptor type, J, protein tyrosine phosphatase, receptor type, J polypeptide, Protein-tyrosine phosphatase eta, Protein-tyrosine phosphatase receptor type J, receptor-type tyrosine-protein phosphatase eta, R-PTP-eta, R-PTP-J, SCC1, susceptibility to colon cancer 1, mouse, homolog of | |
Mouse | |
250 kDa | |
0.1 mg | |
Protein Phosphatase | |
5795 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction