Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DEPDC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570206
Description
This target displays homology in the following species: Cow: 86%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 92%; Rabbit: 86%; Rat: 93%.
May be involved in transcriptional regulation as a transcriptional corepressor. The DEPDC1A-ZNF224 complex may play a critical role in bladder carcinogenesis by repressing the transcription of the A20 gene, leading to transport of NF-KB protein into the nucleus, resulting in suppression of apoptosis of bladder cancer cells.
Specifications
DEPDC1 | |
Polyclonal | |
Unconjugated | |
DEPDC1 | |
5830484J08Rik; cell cycle control protein SDP35; DEP domain containing 1; DEP domain containing 1a; DEP domain-containing protein 1A; DEP.8; Depdc1; DEPDC1A; DEPDC1-V2; SDP35 | |
Rabbit | |
Affinity chromatography | |
RUO | |
55635 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5TB30 | |
DEPDC1 | |
synthetic peptide directed towards the middle region of human DEPDC1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction