Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEPDC1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987311
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7311 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7311 Supplier Bioss Supplier No. BS6525R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May be involved in transcriptional regulation as a transcriptional corepressor. The DEPDC1A-ZNF224 complex may play a critical role in bladder carcinogenesis by repressing the transcription of the A20 gene, leading to transport of NF-KB protein into the nucleus, resulting in suppression of apoptosis of bladder cancer cells.

Specifications

Antigen DEPDC1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DEPDC1
Gene Accession No. Q5TB30
Gene Alias 5830484J08Rik; cell cycle control protein SDP35; DEP domain containing 1; DEP domain containing 1a; DEP domain-containing protein 1A; DEP.8; DEPDC1; DEPDC1A; DEPDC1-V2; LOC100341626; SDP35
Gene Symbols DEPDC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DEPDC1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55635
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.