Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DEPDC1B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572875
Description
DEPDC1B Polyclonal Antibody for Western Blot, IHC (P), ELISA
DEPDC1B is a protein coding gene. Gene Ontology (GO) annotations related to this gene include intracellular signal transduction.
Specifications
DEPDC1B | |
Polyclonal | |
Unconjugated | |
DEPDC1B | |
9830132O03; AW260467; BRCC3; breast cancer cell 3; DEP domain containing 1B; DEP domain-containing protein 1B; DEPDC1B; HBV XAg-transactivated protein 8; HBV X-transactivated gene 8 protein; HBxAg transactivated protein 1; XTP1; XTP8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55789 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q8WUY9 | |
DEPDC1B | |
DEPDC1B antibody for a 19 amino acid peptide near the carboxy terminus of human DEPDC1B. The immunogen is within the last 50 amino acids of DEPDC1B. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction