Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DEPDC1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113277
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113277 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113277 Supplier Invitrogen™ Supplier No. PA5113277
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DEPDC1B Polyclonal Antibody for Western Blot, IHC (P), ICC/IF, ELISA

DEPDC1B is a protein coding gene. Gene Ontology (GO) annotations related to this gene include intracellular signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEPDC1B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene DEPDC1B
Gene Accession No. Q8WUY9
Gene Alias 9830132O03; AW260467; BRCC3; breast cancer cell 3; DEP domain containing 1B; DEP domain-containing protein 1B; DEPDC1B; HBV XAg-transactivated protein 8; HBV X-transactivated gene 8 protein; HBxAg transactivated protein 1; XTP1; XTP8
Gene Symbols DEPDC1B
Host Species Rabbit
Immunogen Recombinant Human DEP domain-containing protein 1B protein (343-529AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55789
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.