Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DEPDC1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567248
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567248 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567248 Supplier Invitrogen™ Supplier No. PA567248
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IGEVPACRLVH RRQLTEANVE EIWKSMTLSY LQKILGLDSL EEVLDVKLVN SKFIIHNVYS VSKQGVVILD DKSKELPHW Highest antigen sequence identity to the following orthologs - mouse 95%, rat 93%.

DEPDC1B is a protein coding gene. Gene Ontology (GO) annotations related to this gene include intracellular signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEPDC1B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DEPDC1B
Gene Accession No. Q8WUY9
Gene Alias 9830132O03; AW260467; BRCC3; breast cancer cell 3; DEP domain containing 1B; DEP domain-containing protein 1B; DEPDC1B; HBV XAg-transactivated protein 8; HBV X-transactivated gene 8 protein; HBxAg transactivated protein 1; XTP1; XTP8
Gene Symbols DEPDC1B
Host Species Rabbit
Immunogen Recombinant Human DEPDC1B. Recombinant protein control fragment (Product #RP-105566).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55789
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.