Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DEPDC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563150
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563150 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563150 Supplier Invitrogen™ Supplier No. PA563150
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YDAQVFRLPG PSRAQCLTTC RSVRERESHS RKSASSCDVS SSPSLPSRTL PTEEVRSQAS DDSSLGKSAN ILMIPHPHLH QYEVSSS Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.

This gene encodes a member of the IML1 family of proteins involved in G-protein signaling pathways. The mechanistic target of rapamycin complex 1 (mTORC1) pathway regulates cell growth by sensing the availability of nutrients. The protein encoded by this gene is a component of the GATOR1 (GAP activity toward Rags) complex which inhibits the amino acid-sensing branch of the mTORC1 pathway. Mutations in this gene are associated with autosomal dominant familial focal epilepsy with variable foci. A single nucleotide polymorphism in an intron of this gene has been associated with an increased risk of hepatocellular carcinoma in individuals with chronic hepatitis C virus infection. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEPDC5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DEPDC5
Gene Accession No. O75140
Gene Alias DEP domain containing 5; DEP domain-containing protein 5; DEP.5; DEPD5; DEPDC5; FFEVF; GATOR complex protein DEPDC5; KIAA0645
Gene Symbols DEPDC5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DEPDC5. Recombinant protein control fragment (Product #RP-95351).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9681
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.