Learn More
Invitrogen™ DEPDC7 Monoclonal Antibody (4D9)

Description
Peptide Sequence: NPSEFKLQKE SDNRMVVKRI FSKAIVDNKN LSKGKTDLLV LFLMDHQKDV FKIPGTLHKI VSVKLMAIQN GRDPNRDAGY IYCQRIDQRD YSNNTEKTTK DELLNLL.
Specifications
Specifications
| Antigen | DEPDC7 |
| Applications | ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 4D9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | DEPDC7 |
| Gene Accession No. | Q96QD5 |
| Gene Alias | AV216087; DEP domain containing 7; DEP domain-containing protein 7; Depdc7; dJ85M6.4; dJ85M6.4 (novel 58.3 KDA protein); novel 58.3 KDA protein; Protein TR2/D15; RGD1309720; TR2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.