Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DEPDC7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569781
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569781 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569781 Supplier Invitrogen™ Supplier No. PA569781
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 92%; Dog: 100%; Guinea Pig: 91%; Horse: 92%; Human: 100%; Mouse: 100%; Pig: 92%; Rabbit: 92%; Rat: 100%.

DEPDC7 gene ontology annotations related to this gene include intracellular signal transduction; regulation of small GTPase mediated signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEPDC7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DEPDC7
Gene Accession No. Q96QD5
Gene Alias AV216087; DEP domain containing 7; DEP domain-containing protein 7; Depdc7; dJ85M6.4; dJ85M6.4 (novel 58.3 KDA protein); novel 58.3 KDA protein; Protein TR2/D15; RGD1309720; TR2
Gene Symbols DEPDC7
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human DEPDC7.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91614
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.