Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DEPDC7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110808
Description
Immunogen sequence: TTIPNQDSQL GKENKLYSPA RYADALFKSS DIRSASLEDL WENLSLKPAN SPHVNISATL SPQVINEVWQ EETIGRLLQL VDLPLLDSLL KQQEAVPKIP QPKRQSTMVN SS Highest antigen sequence indentity to the following orthologs: Mouse - 80%, Rat - 82%.
DEPDC7 gene ontology annotations related to this gene include intracellular signal transduction; regulation of small GTPase mediated signal transduction.
Specifications
| DEPDC7 | |
| Polyclonal | |
| Unconjugated | |
| DEPDC7 | |
| AV216087; DEP domain containing 7; DEP domain-containing protein 7; Depdc7; dJ85M6.4; dJ85M6.4 (novel 58.3 KDA protein); novel 58.3 KDA protein; Protein TR2/D15; RGD1309720; TR2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 91614 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96QD5 | |
| DEPDC7 | |
| Recombinant Protein corresonding to Human DEPDC7. Recombinant protein control fragment (Product #RP-91621). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction