Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DEPP Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 258331AP150UL

Description
DEPP, also named as FIG and C10orf10, is known to be differentially regulated in mouse tissues in response to hypoxia and oxidative stress.Specifications
DEPP | |
Polyclonal | |
Unconjugated | |
DEPP1 | |
C10orf10, DEPP, Fasting induced gene protein, FIG, Protein DEPP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11067 | |
-20°C | |
Liquid |
Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NTK1 | |
DEPP1 | |
DEPP Fusion Protein Ag22773 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction